Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dicyclohexylbenzene 98%

Supplier: AMBEED, INC
Warning

A858661-1G A858661-25G A858661-5G
77199-974EA 9 USD InStock
77199-974 77199-976 77199-978
1,4-Dicyclohexylbenzene 98%
1,4-Dicyclohexylbenzene
Precaución: for Research Use Only


Formula: C₁₈H₂₆
MDL Number: MFCD00001452
CAS Number: 1087-02-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty