Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4-Trimethyl-5-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A862285-1G A862285-25G A862285-5G
77199-992EA 20 USD InStock
77199-992 77199-994 77199-996
1,2,4-Trimethyl-5-nitrobenzene 98%
1,2,4-Trimethyl-5-nitrobenzene
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MDL Number: MFCD00024272
CAS Number: 610-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty