Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-methylpicolinonitrile 97%

Supplier: AMBEED, INC
Warning

A863336-1G A863336-250MG A863336-5G
77199-998EA 145 USD InStock
77199-998 77200-032 77200-034
3-Amino-4-methylpicolinonitrile 97%
3-Amino-4-methylpicolinonitrile
Precaución: for Research Use Only


Formula: C₇H₇N₃
MDL Number: MFCD18384348
CAS Number: 183428-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty