Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dichlorophthalic anhydride 95%

Supplier: AMBEED, INC
Warning

A865510-10G A865510-1G A865510-25G A865510-5G
77200-046EA 45 USD InStock
77200-046 77200-048 77200-050 77200-052
4,5-Dichlorophthalic anhydride 95%
4,5-Dichlorophthalic anhydride
Precaución: for Research Use Only


Formula: C₈H₂Cl₂O₃
MW: 217.01 g/mol
MDL Number: MFCD00075034
CAS Number: 942-06-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty