Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Methoxyphenyl)-1H-imidazole-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A866307-100MG A866307-1G A866307-250MG
77200-054EA 126 USD InStock
77200-054 77200-056 77200-058
1-(2-Methoxyphenyl)-1H-imidazole-5-carboxylic acid 97%
1-(2-Methoxyphenyl)-1H-imidazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O₃
MDL Number: MFCD25368751
CAS Number: 762240-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty