Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoethyl-4-(tert-butyl)phenyl ether 98%

Supplier: AMBEED, INC
Warning

A870462-1G A870462-250MG A870462-5G
77200-082EA 53 USD InStock
77200-082 77200-084 77200-086
2-Bromoethyl-4-(tert-butyl)phenyl ether 98%
2-Bromoethyl-4-(tert-butyl)phenyl ether
Precaución: for Research Use Only


Formula: C₁₂H₁₇BrO
MDL Number: MFCD02030708
CAS Number: 5952-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty