Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ASP-3026 98+%

Supplier: AMBEED, INC
Warning

A874603-10MG A874603-1MG A874603-50MG A874603-5MG
77200-100EA 40 USD InStock
77200-100 77200-102 77200-104 77200-106
ASP-3026 98+%
ASP-3026
Precaución: for Research Use Only


Formula: C₂₉H₄₀N₈O₃S
MDL Number: MFCD21609265
CAS Number: 1097917-15-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty