Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Maleimidopropionic acid hydrazide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A877784-100MG A877784-250MG
77200-126EA 53 USD InStock
77200-126 77200-128
Maleimidopropionic acid hydrazide hydrochloride 95%
Maleimidopropionic acid hydrazide hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉N₃O₃·ClH
MDL Number: MFCD00467760
CAS Number: 293298-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty