Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Piperidylpropiophenone hydrochloride 95%

Supplier: AMBEED, INC
Warning

A886748-1G A886748-250MG A886748-5G
77200-156EA 187 USD InStock
77200-156 77200-158 77200-160
3-Piperidylpropiophenone hydrochloride 95%
3-Piperidylpropiophenone hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₀ClNO
MDL Number: MFCD00035385
CAS Number: 886-06-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty