Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrole-2,3-dicarboxylic acid dimethyl ester 95%

Supplier: AMBEED, INC
Warning

A897080-100MG A897080-1G
77171-822EA 80 USD InStock
77171-822 77171-824
1H-Pyrrole-2,3-dicarboxylic acid dimethyl ester 95%
1H-Pyrrole-2,3-dicarboxylic acid dimethyl ester
Precaución: for Research Use Only


Formula: C₈H₉NO₄
MDL Number: MFCD02328549
CAS Number: 2818-08-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty