Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hesperadin 98%

Supplier: AMBEED, INC
Warning

A899555-10MG A899555-1MG A899555-25MG A899555-50MG
77171-850EA 84 USD InStock
77171-850 77171-852 77171-854 77171-856
Hesperadin 98%
Hesperadin
Precaución: for Research Use Only


Formula: C₂₉H₃₂N₄O₃S
MDL Number: MFCD18074526
CAS Number: 422513-13-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty