Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate 98%

Supplier: AMBEED, INC
Warning

A921862-100MG A921862-1G A921862-250MG
77200-284EA 8 USD InStock
77200-284 77200-286 77200-288
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate 98%
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₂₃H₃₃BF₄N₂
MDL Number: MFCD04973311
CAS Number: 286014-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty