Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzylethylenediamine 98%

Supplier: AMBEED, INC
Danger

A954690-100G A954690-10G A954690-1G A954690-25G A954690-500G A954690-5G
77172-004EA 231 USD InStock
77172-004 77172-006 77172-008 77172-010 77172-012 77172-014
N-Benzylethylenediamine 98%
N-Benzylethylenediamine
Precaución: for Research Use Only


Formula: C₉H₁₄N₂
MDL Number: MFCD00041896
CAS Number: 4152-09-4
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty