Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,4S)-Methyl-4-fluoropyrrolidine-2-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A966862-100MG A966862-1G A966862-250MG A966862-5G
77172-044EA 33 USD InStock
77172-044 77172-046 77172-048 77172-050
(2S,4S)-Methyl-4-fluoropyrrolidine-2-carboxylate hydrochloride 95%
(2S,4S)-Methyl-4-fluoropyrrolidine-2-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₁ClFNO₂
MDL Number: MFCD07374353
CAS Number: 58281-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty