Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-hydroxy-4-nitrobenzoate 95%

Supplier: AMBEED, INC
Warning

A975376-100MG A975376-1G A975376-250MG A975376-5G
77172-080EA 10 USD InStock
77172-080 77172-082 77172-084 77172-086
Methyl 2-hydroxy-4-nitrobenzoate 95%
Methyl 2-hydroxy-4-nitrobenzoate
Precaución: for Research Use Only


Formula: C₈H₇NO₅
MW: 197.145 g/mol
MDL Number: MFCD11041295
CAS Number: 13684-28-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty