Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline 95%

Supplier: AMBEED, INC
Warning

A994065-25G A994065-5G A994065-100G
77200-524EA 101 USD InStock
77200-524 77200-526 77747-744
2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline 95%
2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline
Precaución: for Research Use Only


Formula: C₁₂H₁₇N
MW: 175.27 g/mol
MDL Number: MFCD00957562
CAS Number: 4497-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty