Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Cyanocinnamic acid 98%

Supplier: AMBEED, INC
Warning

A100467-1G A100467-25G A100467-5G
77282-496EA 19 USD InStock
77282-496 77282-498 77282-500
trans-4-Cyanocinnamic acid 98%
trans-4-Cyanocinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₇NO₂
MDL Number: MFCD00016845
CAS Number: 16642-94-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty