Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tucatinib 98%

Supplier: AMBEED, INC
Warning

A100992-10MG A100992-1MG A100992-5MG A100992-100MG A100992-1G A100992-250MG
77245-090EA 17 USD InStock
77245-090 77245-092 77245-094 77745-771 77745-772 77745-773
Tucatinib 98%
Tucatinib
Precaución: for Research Use Only


Formula: C₂₆H₂₄N₈O₂
MW: 480.52 g/mol
MDL Number: MFCD22380467
CAS Number: 937263-43-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty