Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Deoxy-L-ribose 97%

Supplier: AMBEED, INC
Warning

A101449-100G A101449-10G A101449-1G A101449-25G A101449-5G
77282-740EA 714 USD InStock
77282-740 77282-742 77282-744 77282-746 77282-748
2-Deoxy-L-ribose 97%
2-Deoxy-L-ribose
Precaución: for Research Use Only


Formula: C₅H₁₀O₄
MDL Number: MFCD00132941
CAS Number: 18546-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty