Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl ((S)-1-((R)-oxiran-2-yl)-2-phenylethyl)carbamate 98%

Supplier: AMBEED, INC
Warning

A101909-10G A101909-5G
77270-272EA 22 USD InStock
77270-272 77270-274
tert-Butyl ((S)-1-((R)-oxiran-2-yl)-2-phenylethyl)carbamate 98%
tert-Butyl ((S)-1-((R)-oxiran-2-yl)-2-phenylethyl)carbamate
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₃
MDL Number: MFCD00671705
CAS Number: 98760-08-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty