Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

UMI-77 97%

Supplier: AMBEED, INC
Warning

A102389-10MG A102389-1MG A102389-50MG A102389-5MG
77233-122EA 134 USD InStock
77233-122 77233-124 77233-126 77233-128
UMI-77 97%
UMI-77
Precaución: for Research Use Only


Formula: C₁₈H₁₄BrNO₅S₂
MW: 468.34 g/mol
MDL Number: MFCD03471890
CAS Number: 518303-20-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty