Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apoptozole 98%

Supplier: AMBEED, INC
Warning

A102708-10MG A102708-1MG A102708-50MG A102708-5MG
77257-294EA 33 USD InStock
77257-294 77257-296 77257-298 77257-300
Apoptozole 98%
Apoptozole
Precaución: for Research Use Only


Formula: C₃₃H₂₅F₆N₃O₃
MW: 625.60 g/mol
MDL Number: MFCD19443178
CAS Number: 1054543-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty