Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetamidopiperidine 97%

Supplier: AMBEED, INC
Warning

A103587-10G A103587-1G A103587-250MG A103587-25G
77283-120EA 129 USD InStock
77283-120 77283-122 77283-124 77283-126
4-Acetamidopiperidine 97%
4-Acetamidopiperidine
Precaución: for Research Use Only


Formula: C₇H₁₄N₂O
MDL Number: MFCD05254805
CAS Number: 5810-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty