Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylbenzyl acetate 97%

Supplier: AMBEED, INC
Warning

A103727-100G A103727-10G A103727-25G A103727-5G
77283-148EA 113 USD InStock
77283-148 77283-150 77283-152 77283-154
4-Methylbenzyl acetate 97%
4-Methylbenzyl acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
MDL Number: MFCD00017234
CAS Number: 2216-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty