Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N9-Bis(2-hydroxyethyl)nonanediamide 98%

Supplier: AMBEED, INC
Warning

A103800-1MG A103800-25MG
77233-160EA 5 USD InStock
77233-160 77233-162
N1,N9-Bis(2-hydroxyethyl)nonanediamide 98%
N1,N9-Bis(2-hydroxyethyl)nonanediamide
Precaución: for Research Use Only


Formula: C₁₃H₂₆N₂O₄
MDL Number: MFCD00868853
CAS Number: 1675-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty