Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Aminoethyl)-4-methylbenzenesulfonamide 95%

Supplier: AMBEED, INC
Warning

A103957-10G A103957-1G A103957-250MG A103957-25G A103957-5G
77245-280EA 62 USD InStock
77245-280 77245-282 77245-284 77245-286 77245-288
N-(2-Aminoethyl)-4-methylbenzenesulfonamide 95%
N-(2-Aminoethyl)-4-methylbenzenesulfonamide
Precaución: for Research Use Only


Formula: C₉H₁₄N₂O₂S
MW: 214.28 g/mol
MDL Number: MFCD00089604
CAS Number: 14316-16-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty