Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Dioxotetrahydrothiopyran-4-amine 95%

Supplier: AMBEED, INC
Danger

A104032-100MG A104032-1G A104032-250MG A104032-5G
77283-182EA 15 USD InStock
77283-182 77283-184 77283-186 77283-188
1,1-Dioxotetrahydrothiopyran-4-amine 95%
1,1-Dioxotetrahydrothiopyran-4-amine
Precaución: for Research Use Only


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD09540491
CAS Number: 210240-20-3
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty