Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitroisophthalic acid 98%

Supplier: AMBEED, INC
Warning

A104572-100G A104572-10G A104572-1G A104572-5G
77283-244EA 499 USD InStock
77283-244 77283-246 77283-248 77283-250
4-Nitroisophthalic acid 98%
4-Nitroisophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅NO₆
MW: 211.13 g/mol
MDL Number: MFCD00096224
CAS Number: 4315-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty