Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethoxy]phthalazine 98%

Supplier: AMBEED, INC
Warning

A104763-100MG A104763-1G A104763-250MG
77245-340EA 78 USD InStock
77245-340 77245-342 77245-344
(R)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethoxy]phthalazine 98%
(R)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethoxy]phthalazine
Precaución: for Research Use Only


Formula: C₃₈H₂₉N₂OP
MDL Number: MFCD11973799
CAS Number: 828927-95-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty