Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A104997-100G A104997-10G A104997-1G A104997-25G
77283-324EA 198 USD InStock
77283-324 77283-326 77283-328 77283-330
4-Bromobenzylamine hydrochloride 95%
4-Bromobenzylamine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉BrClN
MW: 222.52 g/mol
MDL Number: MFCD00012860
CAS Number: 26177-44-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty