Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EAI-045 97%

Supplier: AMBEED, INC
Warning

A105328-10MG A105328-1G A105328-1MG A105328-250MG A105328-25MG A105328-50MG A105328-5G A105328-5MG
77245-370EA 11 USD InStock
77245-370 77245-372 77245-374 77245-376 77245-378 77245-380 77245-382 77245-384
EAI-045 97%
EAI-045
Precaución: for Research Use Only


Formula: C₁₉H₁₄FN₃O₃S
MDL Number: MFCD30187871
CAS Number: 1942114-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty