Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoropyrimidine 98%

Supplier: AMBEED, INC
Warning

A106096-100MG A106096-1G A106096-250MG
77257-440EA 9 USD InStock
77257-440 77549-032 77747-789
2-Fluoropyrimidine 98%
2-Fluoropyrimidine
Precaución: for Research Use Only


Formula: C₄H₃FN₂
MDL Number: MFCD08059530
CAS Number: 31575-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty