Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenyl-2-methyl-3-butyn-2-ol 97%

Supplier: AMBEED, INC
Warning

A106437-100G A106437-1G A106437-250MG A106437-25G A106437-5G
77270-564EA 455 USD InStock
77270-564 77270-566 77270-568 77270-570 77270-572
4-Phenyl-2-methyl-3-butyn-2-ol 97%
4-Phenyl-2-methyl-3-butyn-2-ol
Precaución: for Research Use Only


Formula: C₁₁H₁₂O
MDL Number: MFCD00041572
CAS Number: 1719-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty