Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A106810-100G A106810-5G
77257-460EA 698 USD InStock
77257-460 77257-462
1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine hydrochloride 97%
1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₅ClN₂S
MDL Number: MFCD07369525
CAS Number: 908591-25-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty