Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-methoxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A106844-100MG A106844-1G A106844-250MG A106844-25G A106844-5G
77270-610EA 5 USD InStock
77270-610 77270-612 77270-614 77270-616 77270-618
3-Chloro-5-methoxybenzaldehyde 97%
3-Chloro-5-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
MDL Number: MFCD08234653
CAS Number: 164650-68-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty