Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Methylbenzylidene)camphor 98%

Supplier: AMBEED, INC
Danger

A106890-1G A106890-5G A106890-25G
77270-620EA 6 USD InStock
77270-620 77270-622 77747-795
3-(4-Methylbenzylidene)camphor 98%
3-(4-Methylbenzylidene)camphor
Precaución: for Research Use Only


Formula: C₁₈H₂₂O
MW: 254.37 g/mol
MDL Number: MFCD00209662
CAS Number: 36861-47-9
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty