Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Hydroxyethoxy)benzaldehyde 97%

Supplier: AMBEED, INC
Warning

A107227-100G A107227-10G A107227-1G A107227-250MG A107227-5G
77245-490EA 494.99 USD InStock
77245-490 77245-492 77245-494 77245-496 77745-804
4-(2-Hydroxyethoxy)benzaldehyde 97%
4-(2-Hydroxyethoxy)benzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MDL Number: MFCD00191450
CAS Number: 22042-73-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty