Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,3-difluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A107442-10G A107442-1G A107442-250MG A107442-25G A107442-5G
77283-600EA 95 USD InStock
77283-600 77283-602 77283-604 77283-606 77283-608
4-Chloro-2,3-difluorobenzoic acid 97%
4-Chloro-2,3-difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃ClF₂O₂
MW: 192.546 g/mol
MDL Number: MFCD07368122
CAS Number: 150444-94-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty