Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pentynoic acid 95%

Supplier: AMBEED, INC
Warning

A107561-10G A107561-1G A107561-250MG A107561-25G A107561-5G
77270-668EA 128 USD InStock
77270-668 77270-670 77270-672 77270-674 77270-676
2-Pentynoic acid 95%
2-Pentynoic acid
Precaución: for Research Use Only


Formula: C₅H₆O₂
MDL Number: MFCD00014370
CAS Number: 5963-77-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty