Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-methylnicotinate 98%

Supplier: AMBEED, INC
Warning

A107977-100G A107977-10G A107977-1G A107977-25G A107977-5G
77245-534EA 195 USD InStock
77245-534 77245-536 77245-538 77245-540 77245-542
Ethyl-2-methylnicotinate 98%
Ethyl 2-methylnicotinate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00006336
CAS Number: 1721-26-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty