Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Andrographolide 98+%

Supplier: AMBEED, INC
Warning

A108018-100G A108018-100MG A108018-1G A108018-25G A108018-500G A108018-5G
77283-674EA 283 USD InStock
77283-674 77283-676 77283-678 77283-680 77283-682 77283-684
Andrographolide 98+%
Andrographolide
Precaución: for Research Use Only


Formula: C₂₀H₃₀O₅
MDL Number: MFCD07778082
CAS Number: 5508-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty