Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluorophenol 97%

Supplier: AMBEED, INC
Warning

A108452-100G A108452-10G A108452-1G A108452-25G A108452-5G
77283-736EA 243 USD InStock
77283-736 77283-738 77283-740 77283-742 77283-744
5-Chloro-2-fluorophenol 97%
5-Chloro-2-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₄ClFO
MW: 146.55
MDL Number: MFCD04116357
CAS Number: 186589-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty