Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-nitrobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A108736-250MG A108736-5G A108736-1G
77270-724EA 11 USD InStock
77270-724 77270-726 77747-800
3-Chloro-5-nitrobenzoic acid 98%
3-Chloro-5-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD09263224
CAS Number: 34662-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty