Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,4-Difluorophenyl)hydrazine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A108848-100G A108848-1G A108848-25G A108848-5G
77283-780EA 139 USD InStock
77283-780 77283-782 77283-784 77283-786
(3,4-Difluorophenyl)hydrazine hydrochloride 98%
(3,4-Difluorophenyl)hydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇ClF₂N₂
MW: 213.49
MDL Number: MFCD03094170
CAS Number: 875664-54-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty