Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Isopropylimidazole 97%

Supplier: AMBEED, INC
Warning

A109006-100G A109006-10G A109006-25G
77233-396EA 21 USD InStock
77233-396 77233-398 77233-400
1-Isopropylimidazole 97%
1-Isopropylimidazole
Precaución: for Research Use Only


Formula: C₆H₁₀N₂
MDL Number: MFCD00234239
CAS Number: 4532-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty