Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-formylbenzenesulfonic acid 97%

Supplier: AMBEED, INC
Danger

A109348-10G A109348-5G
77257-592EA 134 USD InStock
77257-592 77257-594
4-Chloro-3-formylbenzenesulfonic acid 97%
4-Chloro-3-formylbenzenesulfonic acid
Precaución: for Research Use Only


Formula: C₇H₅ClO₄S
MW: 220.63 g/mol
MDL Number: MFCD30717632
CAS Number: 60767-69-3
UN: 2585
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty