Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-pyridineboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A110138-1G A110138-250MG A110138-5G
77257-624EA 22 USD InStock
77257-624 77257-626 77257-628
2-Benzyloxy-5-pyridineboronic acid pinacol ester 95%
2-Benzyloxy-5-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₈H₂₂BNO₃
MW: 311.19 g/mol
MDL Number: MFCD06798263
CAS Number: 832735-54-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty