Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-α-Phenylglycine 98%

Supplier: AMBEED, INC
Warning

A110145-1000G A110145-100G A110145-25G A110145-500G A110145-10G
77245-658EA 93 USD InStock
77245-658 77245-660 77245-662 77245-664 77614-844
L(+)-α-Phenylglycine 98%
L(+)-α-Phenylglycine
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00064403
CAS Number: 2935-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty