Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Difluoromethoxy)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A110170-1G A110170-25G A110170-5G
77233-500EA 9 USD InStock
77233-500 77233-502 77233-504
2-(Difluoromethoxy)benzoic acid 98%
2-(Difluoromethoxy)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₆F₂O₃
MDL Number: MFCD00236223
CAS Number: 97914-59-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty