Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-acetamido-5-chloro-2-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A110465-10G A110465-1G A110465-25G A110465-5G
77284-002EA 45 USD InStock
77284-002 77284-004 77284-006 77284-008
Methyl-4-acetamido-5-chloro-2-hydroxybenzoate 98%
Methyl-4-acetamido-5-chloro-2-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀ClNO₄
MW: 243.6400 g/mol
MDL Number: MFCD02093078
CAS Number: 24190-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty